Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053318.1 | NP_445770.1 | 9 | 1032 | ||
GenBank mRNA | BC091137.1 | - | 9 | 1023 | 7326169 | |
FQ210231.1 | - | 9 | 1034 | |||
FQ211176.1 | - | 9 | 1034 | |||
FQ218633.1 | - | 9 | 1031 | |||
FQ218971.1 | - | 9 | 1022 | |||
FQ218998.1 | - | 6 | 801 | |||
FQ219255.1 | - | 4 | 481 |