Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175754.1 | NP_786930.1 | 3 | 447 | ||
XM_006239541.2 | XP_006239603.1 | 4 | 652 | |||
XM_006239542.2 | XP_006239604.1 | 4 | 652 | |||
XM_008764338.1 | XP_008762560.1 | 4 | 652 | |||
XM_008764339.1 | XP_008762561.1 | 4 | 652 | |||
XM_008764340.1 | XP_008762562.1 | 3 | 347 | |||
GenBank mRNA | M64780.1 | - | 3 | 447 |