Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012496.2 | NP_036628.2 | 3 | 248 | ||
GenBank mRNA | BC081697.1 | - | 3 | 248 | 7109764 | |
FQ209505.1 | - | 3 | 268 | |||
FQ209538.1 | - | 3 | 268 | |||
FQ209583.1 | - | 3 | 268 | |||
FQ209594.1 | - | 3 | 268 | |||
FQ209615.1 | - | 3 | 268 | |||
FQ209621.1 | - | 3 | 266 | |||
FQ209639.1 | - | 3 | 268 | |||
FQ209643.1 | - | 3 | 265 | |||
FQ209782.1 | - | 3 | 268 | |||
FQ209819.1 | - | 3 | 278 | |||
FQ210002.1 | - | 3 | 268 | |||
FQ211083.1 | - | 3 | 268 | |||
FQ211156.1 | - | 3 | 268 | |||
FQ211159.1 | - | 3 | 268 | |||
FQ218139.1 | - | 3 | 268 | |||
FQ218217.1 | - | 3 | 268 | |||
FQ218348.1 | - | 3 | 274 | |||
FQ218402.1 | - | 3 | 270 | |||
FQ218522.1 | - | 3 | 268 | |||
FQ218551.1 | - | 3 | 268 | |||
FQ218665.1 | - | 3 | 268 | |||
FQ218885.1 | - | 3 | 268 | |||
FQ218911.1 | - | 3 | 268 | |||
FQ219116.1 | - | 3 | 268 | |||
FQ219236.1 | - | 3 | 267 | |||
FQ219261.1 | - | 3 | 268 | |||
FQ219453.1 | - | 3 | 268 | |||
FQ219555.1 | - | 3 | 268 | |||
FQ219574.1 | - | 3 | 268 | |||
FQ219585.1 | - | 3 | 268 | |||
FQ219640.1 | - | 3 | 266 | |||
FQ219646.1 | - | 3 | 268 | |||
FQ219703.1 | - | 3 | 267 | |||
M10149.1 | - | 3 | 259 |