Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012515.2 | NP_036647.1 | 4 | 615 | ||
GenBank mRNA | FQ228784.1 | - | 4 | 617 | ||
J05122.1 | - | 3 | 558 |