Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012530.2 | NP_036662.1 | 5 | 614 | ||
GenBank mRNA | DN931989.1 | - | 5 | 613 | ||
FQ214594.1 | - | 5 | 590 | |||
FQ214655.1 | - | 5 | 590 | |||
FQ214750.1 | - | 5 | 590 | |||
FQ214825.1 | - | 4 | 411 | |||
FQ214931.1 | - | 5 | 591 | |||
FQ215369.1 | - | 4 | 488 | |||
FQ215410.1 | - | 5 | 590 | |||
FQ215629.1 | - | 3 | 207 | |||
FQ215927.1 | - | 4 | 443 | |||
FQ216321.1 | - | 4 | 442 | |||
FQ216507.1 | - | 5 | 585 | |||
FQ216611.1 | - | 3 | 207 | |||
FQ217421.1 | - | 4 | 455 | |||
FQ224848.1 | - | 4 | 441 | |||
FQ224918.1 | - | 5 | 591 | |||
M10140.1 | - | 5 | 587 | |||
M14864.1 | - | 3 | 260 |