Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289737.1 | NP_001276666.1 | 4 | 740 | ||
NM_012534.4 | NP_036666.2 | 3 | 716 | |||
XM_008772796.1 | XP_008771018.1 | 4 | 1210 | |||
GenBank mRNA | BC126082.1 | - | 3 | 716 | 8364912 | |
U47922.1 | - | 3 | 669 | |||
V01219.1 | - | 3 | 502 |