Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012551.2 | NP_036683.1 | 2 | 523 | ||
GenBank mRNA | AY551092.1 | - | 2 | 523 | ||
M18416.1 | - | 875 |