Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012559.2 | NP_036691.2 | 3 | 174 | ||
XM_006232525.1 | XP_006232587.1 | 3 | 328 | |||
GenBank mRNA | BC078893.1 | - | 3 | 174 | 7122366 | |
FQ219408.1 | - | 3 | 194 | |||
FQ219511.1 | - | 3 | 196 | |||
FQ219541.1 | - | 3 | 196 | |||
FQ219604.1 | - | 3 | 195 | |||
FQ229132.1 | - | 3 | 201 | |||
J00734.1 | - | 3 | 159 |