Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012600.2 | NP_036732.2 | 7, 8 | 809 | ||
XM_008766439.1 | XP_008764661.1 | 7, 8 | 1106 | |||
GenBank mRNA | CO560778.1 | - | 3, 4 | 306 | ||
FQ228526.1 | - | 7, 8 | 840 |