Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012685.2 | NP_036817.1 | 496 | |||
GenBank mRNA | BC087136.1 | - | 496 | 7124910 | ||
M86835.1 | - | 465 |
Set Membership: |
![]() |