Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012702.2 | NP_036834.2 | 2 | 512 | ||
GenBank mRNA | BC098921.1 | - | 2 | 512 | 7441743 | |
M32474.1 | - | 2 | 497 |