Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012706.2 | NP_036838.1 | 2 | 631 | ||
GenBank mRNA | X56661.1 | - | 2 | 723 |
Set Membership: |
![]() |