Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012730.1 | NP_036862.1 | 5 | 870 | ||
GenBank mRNA | AB008423.1 | - | 5 | 746 | ||
BC078897.1 | - | 5 | 805 | 7109496 | ||
FQ218496.1 | - | 5 | 812 | |||
M16655.1 | - | 5 | 796 | |||
M22330.1 | - | 5 | 870 |