Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012733.4 | NP_036865.1 | 2 | 263 | ||
GenBank mRNA | FQ209770.1 | - | 2 | 277 | ||
FQ209927.1 | - | 2 | 247 | |||
FQ210835.1 | - | 2 | 278 | |||
FQ218090.1 | - | 2 | 277 | |||
M16459.1 | - | 2 | 263 | |||
M19257.1 | - | 2 | 193 |