Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012755.1 | NP_036887.1 | 8 | 916 | ||
XM_006256520.2 | XP_006256582.1 | 6 | 1751 | |||
XM_006256521.2 | XP_006256583.1 | 6 | 1751 | |||
XM_006256522.1 | XP_006256584.1 | 7 | 1203 | |||
GenBank mRNA | FQ232093.1 | - | 1 | 25 | ||
U35365.1 | - | 8 | 916 |