Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012755.1 | NP_036887.1 | 12, 13 | 1511 | ||
XM_006256520.2 | XP_006256582.1 | 10, 11 | 2346 | |||
XM_006256521.2 | XP_006256583.1 | 10, 11 | 2337 | |||
XM_006256522.1 | XP_006256584.1 | 11, 12 | 1798 | |||
GenBank mRNA | FQ231621.1 | - | 3, 4 | 396 | ||
FQ232093.1 | - | 5, 6 | 611 | |||
U35365.1 | - | 12, 13 | 1511 |