Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245672.2 | XP_006245734.1 | 6 | 2774 | ||
XM_006245673.2 | XP_006245735.1 | 6 | 3150 | |||
XM_006245674.2 | XP_006245736.1 | 6 | 3103 | |||
XM_008767418.1 | XP_008765640.1 | 6 | 2727 | |||
XM_008767419.1 | XP_008765641.1 | 4 | 2124 | |||
GenBank mRNA | DQ266367.1 | - | 1 | 217 |