Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016996.1 | NP_058692.1 | 4 | 1105 | ||
GenBank mRNA | AY214122.1 | - | 4 | 1095 | ||
U10354.1 | - | 4 | 1105 | |||
U20289.1 | - | 3 | 731 |
Set Membership: |
![]() |