Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016999.2 | NP_058695.2 | 4 | 463 | ||
XM_008763976.1 | XP_008762198.1 | 4 | 537 | |||
GenBank mRNA | BC074012.1 | - | 4 | 473 | 7100197 | |
M29853.1 | - | 4 | 463 |