Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017031.2 | NP_058727.2 | 2 | 916 | ||
GenBank mRNA | AF202732.1 | - | 774 | |||
AF202733.1 | - | 2 | 480 | |||
U95748.1 | - | 2 | 916 |