Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017065.1 | NP_058761.1 | 4, 5 | 620 | ||
GenBank mRNA | AY742860.1 | - | 950 | |||
X15468.1 | - | 4, 5 | 620 |