Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017068.2 | NP_058764.2 | 3, 4 | 501 | ||
XM_006257486.2 | XP_006257548.1 | 3, 4 | 532 | |||
GenBank mRNA | BC061990.1 | - | 3, 4 | 501 | 5599402 | |
D90211.1 | - | 3, 4 | 485 | |||
FQ211284.1 | - | 3, 4 | 500 | |||
M32016.1 | - | 2, 3 | 339 |