Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017080.2 | NP_058776.2 | 3 | 635 | ||
XM_006250471.2 | XP_006250533.1 | 4 | 416 | |||
XM_006250473.2 | XP_006250535.1 | 4 | 451 | |||
GenBank mRNA | AF542063.1 | - | 2 | 235 | ||
BC078865.1 | - | 3 | 456 | 7109256 | ||
FQ209646.1 | - | 3 | 402 | |||
FQ209829.1 | - | 3 | 402 | |||
FQ210302.1 | - | 3 | 400 | |||
FQ210596.1 | - | 3 | 402 | |||
FQ211059.1 | - | 3 | 399 | |||
FQ211404.1 | - | 3 | 401 | |||
FQ212763.1 | - | 3 | 403 | |||
FQ214152.1 | - | 3 | 402 | |||
FQ214397.1 | - | 4 | 458 | |||
FQ218558.1 | - | 3 | 400 | |||
FQ218584.1 | - | 3 | 402 | |||
FQ219123.1 | - | 3 | 402 | |||
FQ219241.1 | - | 3 | 401 | |||
FQ219385.1 | - | 3 | 400 | |||
FQ219394.1 | - | 3 | 400 | |||
FQ219517.1 | - | 3 | 402 | |||
FQ219567.1 | - | 3 | 411 | |||
FQ219624.1 | - | 3 | 335 | |||
FQ220122.1 | - | 3 | 400 | |||
J05107.1 | - | 3 | 383 |