Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022498.2 | NP_071943.1 | 7 | 1222 | ||
GenBank mRNA | D90165.1 | - | 7 | 1051 | ||
FM122849.1 | - | 4 | 534 | |||
FQ211612.1 | - | 2 | 175 | |||
FQ228186.1 | - | 3 | 336 | |||
FQ232892.1 | - | 3 | 336 |