Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024353.1 | NP_077329.1 | 7, 8 | 644 | ||
XM_006235083.2 | XP_006235145.1 | 7, 8 | 681 | |||
XM_006235085.2 | XP_006235147.1 | 7, 8 | 542 | |||
XM_006235088.2 | XP_006235150.1 | 6, 7 | 2795 | |||
XM_006235089.2 | XP_006235151.1 | 7, 8 | 3368 | |||
XM_006235094.2 | XP_006235156.1 | 7, 8 | 681 | |||
XM_008762248.1 | XP_008760470.1 | 7, 8 | 2914 | |||
XM_008762249.1 | XP_008760471.1 | 7, 8 | 2914 | |||
XM_008762250.1 | XP_008760472.1 | 7, 8 | 2914 | |||
XM_008762251.1 | XP_008760473.1 | 7, 8 | 2914 | |||
GenBank mRNA | AF031370.1 | - | 5, 6 | 414 | ||
L15556.1 | - | 7, 8 | 644 | |||
L18962.1 | - | 7, 8 | 547 | |||
U57836.1 | - | 7, 8 | 547 |