Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033967.2 | NP_001029139.1 | 2 | 168 | ||
NM_030986.2 | NP_112248.2 | 2 | 168 | |||
GenBank mRNA | BC061727.1 | - | 2 | 122 | 5598607 | |
CO570724.1 | - | 2 | 168 | |||
D13061.1 | - | 2 | 135 |