Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130819.1 | NP_570832.1 | 3 | 646 | ||
GenBank mRNA | AF337953.1 | - | 3 | 646 |