Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134432.2 | NP_602308.1 | 2 | 378 | ||
XM_008772597.1 | XP_008770819.1 | 3 | 578 | |||
GenBank mRNA | BC078741.1 | - | 2 | 378 | 7128542 | |
BC087679.1 | - | 1 | 214 | 7318795 |