Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014443.2 | NP_001014443.1 | 8, 9 | 1483 | ||
NM_001319848.1 | NP_001306777.1 | 7, 8 | 1342 | |||
NM_012475.4 | NP_036607.3 | 7, 8 | 1342 | |||
XM_011509414.1 | XP_011507716.1 | 7, 8 | 1405 | |||
XM_017001036.1 | XP_016856525.1 | 7, 8 | 1405 | |||
XM_017001037.1 | XP_016856526.1 | 8, 9 | 1459 | |||
XM_017001038.1 | XP_016856527.1 | 8, 9 | 1495 | |||
XM_017001039.1 | XP_016856528.1 | 7, 8 | 1348 | |||
XM_017001040.1 | XP_016856529.1 | 7, 8 | 1405 | |||
XM_017001041.1 | XP_016856530.1 | 8, 9 | 1144 | |||
XM_017001042.1 | XP_016856531.1 | 3, 4 | 303 | |||
GenBank mRNA | AF177758.1 | - | 7, 8 | 1193 | ||
AF217979.1 | - | 7, 8 | 1266 | |||
AF233442.1 | - | 7, 8 | 1280 | |||
AK292319.1 | - | 8, 9 | 1428 | |||
AK310326.1 | - | 7, 8 | 1375 | |||
AL157417.1 | - | 5, 6 | 1193 | |||
BC003130.1 | - | 6, 7 | 842 | |||
BC090946.1 | - | 8, 9 | 1431 | 3877298 | ||
BC136291.1 | - | 6, 7 | 1130 | 9020278 |