Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271058.1 | NP_001257987.1 | 10 | 1295 | ||
NM_001271059.1 | NP_001257988.1 | 11 | 1339 | |||
NM_001271060.1 | NP_001257989.1 | 10 | 1330 | |||
NM_001271061.1 | NP_001257990.1 | 10 | 1264 | |||
NM_001271062.1 | NP_001257991.1 | 10 | 1373 | |||
NM_173096.3 | NP_775119.2 | 10 | 1453 | |||
XM_006248146.2 | XP_006248208.1 | 11 | 1519 | |||
XM_006248149.2 | XP_006248211.1 | 11 | 1562 | |||
XM_008768555.1 | XP_008766777.1 | 10 | 3455 | |||
XM_008768556.1 | XP_008766778.1 | 10 | 3428 | |||
XM_008768557.1 | XP_008766779.1 | 10 | 3363 | |||
GenBank mRNA | BC099784.1 | - | 10 | 1441 | 7933587 | |
FQ226931.1 | - | 10 | 1330 | |||
FQ230542.1 | - | 10 | 1342 | |||
FQ231581.1 | - | 11 | 1407 | |||
X52711.1 | - | 9 | 1247 |