Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173099.1 | NP_775122.1 | 1 | 511 | ||
XM_006254626.2 | XP_006254688.1 | 2 | 1096 | |||
XM_006254627.2 | XP_006254689.1 | 1 | 1003 | |||
GenBank mRNA | L43592.1 | - | 1 | 511 |
Gene Symbol: Pcdhb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114603.1 | NP_001108075.1 | 1 | 528 |
Gene Symbol: Pcdhb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114601.1 | NP_001108073.1 | 1 | 555 |
Gene Symbol: Pcdhb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014779.1 | NP_001014779.1 | 1 | 374 |