Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013236.3 | NP_037368.1 | 2 | 405 | ||
GenBank mRNA | AF086923.1 | - | 2 | 348 | ||
AF119662.1 | - | 2 | 345 | |||
AK025734.1 | - | 2 | 336 | |||
AK095309.1 | - | 2 | 173 | |||
AL050282.1 | - | 2 | 337 | |||
BC007508.2 | - | 2 | 327 | 3030062 | ||
BI597255.1 | - | 2 | 404 | |||
CR456568.1 | - | 2 | 190 | |||
CR457381.1 | - | 2 | 139 | |||
DB035174.1 | - | 2 | 405 |