Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173311.2 | NP_775433.2 | 2, 3 | 244 | ||
GenBank mRNA | AB092812.1 | - | 2, 3 | 146 | ||
BC097931.1 | - | 2, 3 | 244 | 7389090 |