Search Thermo Fisher Scientific
Gene Symbol: Tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173839.2 | NP_776211.1 | 2 | 271 | ||
XM_006241352.2 | XP_006241414.1 | 2 | 324 | |||
XM_008765357.1 | XP_008763579.1 | 2 | 324 | |||
GenBank mRNA | AY098915.1 | - | 2 | 271 |