Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175844.3 | NP_787038.1 | 2 | 1206 | ||
GenBank mRNA | AF336113.1 | - | 2 | 1206 | ||
BC158575.1 | - | 2 | 1192 | 9033092 |