Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199499.1 | NP_955793.1 | 9 | 1588 | ||
XM_008759169.1 | XP_008757391.1 | 8 | 2031 | |||
XM_008759170.1 | XP_008757392.1 | 9 | 1302 | |||
GenBank mRNA | BC064659.1 | - | 9 | 1588 | 5623799 |