Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr1op2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201421.1 | NP_958824.1 | 3 | 537 | ||
XM_006237688.2 | XP_006237750.1 | 3 | 526 | |||
XM_008763402.1 | XP_008761624.1 | 3 | 516 | |||
GenBank mRNA | AY426739.1 | - | 3 | 537 | ||
AY426740.1 | - | 3 | 537 | |||
BC087696.1 | - | 3 | 434 | 7312267 |