Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201635.2 | NP_963966.1 | 2 | 357 | ||
XM_006235672.2 | XP_006235734.1 | 2 | 699 | |||
GenBank mRNA | BC100059.1 | - | 2 | 702 | 7129117 | |
X59506.1 | - | 2 | 700 |