Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tepp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201655.1 | NP_970617.1 | 4 | 630 | ||
XM_008772274.1 | XP_008770496.1 | 4 | 907 | |||
XM_008772275.1 | XP_008770497.1 | 4 | 907 | |||
XM_008772276.1 | XP_008770498.1 | 4 | 907 |