Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203335.2 | NP_976080.1 | 2, 3 | 303 | ||
GenBank mRNA | AY423047.1 | - | 2, 3 | 286 | ||
BC166413.1 | - | 299 | 9100951 | |||
CB314647.1 | - | 2, 3 | 319 | |||
FQ210919.1 | - | 2, 3 | 345 |