Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203409.1 | NP_981954.1 | 2 | 330 | ||
GenBank mRNA | AY495695.1 | - | 2 | 330 | ||
AY495696.1 | - | 2 | 450 |