Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amigo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206881.1 | NP_996764.1 | 1 | 505 | ||
XM_006233140.2 | XP_006233202.1 | 2 | 905 | |||
XM_008761391.1 | XP_008759613.1 | 2 | 940 | |||
XM_008761392.1 | XP_008759614.1 | 1 | 1362 | |||
GenBank mRNA | AY237729.1 | - | 1 | 505 | ||
BC167749.1 | - | 1 | 549 | 9021924 |