Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207591.1 | NP_997474.1 | 12 | 1400 | ||
GenBank mRNA | AY535002.1 | - | 12 | 1400 | ||
BC086359.1 | - | 12 | 1404 | 7304013 | ||
FQ212888.1 | - | 12 | 1435 | |||
FQ213201.1 | - | 12 | 1409 | |||
FQ215499.1 | - | 10 | 1101 | |||
FQ219058.1 | - | 12 | 1407 | |||
FQ222789.1 | - | 12 | 1410 |