Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psbpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207593.2 | NP_997476.2 | 2 | 137 | ||
GenBank mRNA | BC099748.1 | - | 2 | 131 | ||
J00776.1 | - | 1 | 49 | |||
V01256.1 | - | 2 | 134 |