Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207615.2 | NP_997498.2 | 2 | 518 | ||
GenBank mRNA | AY365006.1 | - | 2 | 265 | ||
BC098643.1 | - | 2 | 518 | 7123848 |