Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000861.3 | NP_000852.1 | 3 | 2580 | ||
NM_001098211.1 | NP_001091681.1 | 2 | 2501 | |||
NM_001098212.1 | NP_001091682.1 | 2 | 2451 | |||
NM_001098213.1 | NP_001091683.1 | 2 | 2731 | |||
XM_011533652.1 | XP_011531954.1 | 4 | 2868 | |||
XM_011533653.2 | XP_011531955.1 | 2 | 2697 | |||
XM_017006283.1 | XP_016861772.1 | 2 | 2651 | |||
XM_017006284.1 | XP_016861773.1 | 2 | 2690 | |||
GenBank mRNA | BC060802.1 | - | 3 | 2580 | 30340368 |
Set Membership: |
![]() |