Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109.4 | NP_001100.3 | 8, 9 | 790 | ||
NM_001164489.1 | NP_001157961.1 | 8, 9 | 790 | |||
NM_001164490.1 | NP_001157962.1 | 7, 8 | 673 | |||
XM_011539117.2 | XP_011537419.2 | 9, 10 | 971 | |||
XM_017015465.1 | XP_016870954.1 | 9, 10 | 971 | |||
XM_017015466.1 | XP_016870955.1 | 9, 10 | 973 | |||
GenBank mRNA | AK301147.1 | - | 7, 8 | 638 | ||
AK304324.1 | - | 8, 9 | 947 | |||
AK310662.1 | - | 7, 8 | 646 | |||
AK312834.1 | - | 8, 9 | 755 | |||
BC064500.1 | - | 8, 9 | 737 | |||
BC115404.1 | - | 8, 9 | 709 | 40075505 | ||
D26579.1 | - | 8, 9 | 713 |