Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256252.1 | NP_001243181.1 | 4 | 385 | ||
NM_001256253.1 | NP_001243182.1 | 5 | 541 | |||
NM_001974.4 | NP_001965.3 | 5 | 541 | |||
XM_011527794.1 | XP_011526096.1 | 5 | 541 | |||
GenBank mRNA | AK131562.1 | - | 4 | 355 | ||
AK290401.1 | - | 5 | 511 | |||
AK313495.1 | - | 5 | 511 | |||
BC059395.1 | - | 5 | 524 | 30349201 | ||
X81479.1 | - | 5 | 519 |
Set Membership: |
G-protein coupled receptor |