Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016540.3 | NP_057624.3 | 4 | 1562 | ||
XM_011542559.2 | XP_011540861.1 | 3 | 1496 | |||
GenBank mRNA | AB040973.1 | - | 4 | 1395 | ||
BC067473.1 | - | 4 | 1390 | 6971838 | ||
BC067474.1 | - | 4 | 1390 | 6971839 |
Set Membership: |
![]() |