Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPK1 SIRNA Assays ›
Gene Symbol:
TPK1
Gene Name:
thiamin pyrophosphokinase 1
Gene Aliases:
HTPK1, PP20, THMD5
Chromosome Location:
Chr.7: 144451941 - 144836424 on Build GRCh38
UniGene ID:
Hs.660232
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
202139
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042482.1
NP_001035947.1
8
1203
NM_022445.3
NP_071890.2
9
1350
XM_005249970.1
XP_005250027.1
9
1328
XM_011516031.1
XP_011514333.1
10
1429
XM_011516032.2
XP_011514334.1
10
1565
XM_011516033.2
XP_011514335.1
10
1435
XM_011516034.2
XP_011514336.1
10
1492
XM_011516037.2
XP_011514339.1
9
1676
XM_011516039.2
XP_011514341.1
9
1611
XM_011516041.2
XP_011514343.1
8
1579
XM_011516043.1
XP_011514345.1
9
1282
XM_011516044.1
XP_011514346.1
6
1106
XM_011516047.2
XP_011514349.1
6
2100
XM_011516048.1
XP_011514350.1
5
1066
XM_011516050.1
XP_011514352.1
7
1159
XM_017011970.1
XP_016867459.1
9
1403
XM_017011971.1
XP_016867460.1
9
1353
XM_017011973.1
XP_016867462.1
9
1495
XM_017011974.1
XP_016867463.1
9
1340
XM_017011975.1
XP_016867464.1
9
1291
XM_017011976.1
XP_016867465.1
10
1456
XM_017011977.1
XP_016867466.1
10
1443
XM_017011978.1
XP_016867467.1
9
1402
XM_017011979.1
XP_016867468.1
8
1335
XM_017011980.1
XP_016867469.1
6
1188
XM_017011981.1
XP_016867470.1
5
1282
XM_017011982.1
XP_016867471.1
7
1249
XM_017011983.1
XP_016867472.1
7
1193
XM_017011984.1
XP_016867473.1
7
1252
XM_017011985.1
XP_016867474.1
7
1208
XM_017011986.1
XP_016867475.1
7
1214
GenBank mRNA
AB028138.1
-
8
1265
AF297710.1
-
9
1337
AK091736.1
-
10
1411
AK128661.1
-
7
1156
AK289652.1
-
9
1325
AK308693.1
-
10
1472
AY206415.1
-
9
1332
BC040555.1
-
12
1726
5269277
BC068460.1
-
9
1329
5288626
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
thiamine metabolic process
thiamine diphosphate biosynthetic process
phosphorylation
thiamine-containing compound metabolic process
thiamine diphosphokinase activity
ATP binding
kinase activity
thiamine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0